accessibility menu, dialog, popup

UserName

Viewing profile as user:
SiSiCa

CAS RN 12013-56-8

IUPAC Name: calcium bis(λ⁻¹-silanuide)
Synonyms: calcium bis(λ⁻¹-silanuide)
Molecular Weight (g/mol): 96.25
Molecular Formula: CaSi2
InChi Key: XXCKHUMTXOCLPT-UHFFFAOYSA-N
SMILES: [Si-].[Si-].[Ca++]
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results