accessibility menu, dialog, popup

UserName

Viewing profile as user:
CeOO

CAS RN 12014-56-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 212.18
Molecular Formula: CeH8O4
InChi Key: DPUCLPLBKVSJIB-UHFFFAOYSA-N
SMILES: O.O.O.O.[Ce]
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)