accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12014-85-6

CeSiSi

CAS RN 12014-85-6

IUPAC Name: (silanylidenecerio)silane
Synonyms: (silanylidenecerio)silane
Molecular Weight (g/mol): 200.32
Molecular Formula: CeH4Si2
InChi Key: YARAWHILSWOVAK-UHFFFAOYSA-N
SMILES: [SiH2]=[Ce]=[SiH2]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results