accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNNCH3OH3CFF(E/Z)

CAS RN 1201643-08-4

IUPAC Name: 2,2-difluoro-6-methyl-4-[2-(4-propoxyphenyl)ethenyl]-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Synonyms: 2,2-difluoro-6-methyl-4-[2-(4-propoxyphenyl)ethenyl]-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Molecular Weight (g/mol): 366.22
Molecular Formula: C21H21BF2N2O
InChi Key: NDQRWYKJLFZGBK-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(C=CC2=CC(C)=C3C=C4C=CC=[N+]4[B-](F)(F)N23)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results