accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12017-01-5

IUPAC Name: titanium(4+) λ²-cobalt(2+) trioxidandiide
Synonyms: cobalt titanium oxide
Molecular Weight (g/mol): 154.8
Molecular Formula: CoO3Ti
InChi Key: LFSBSHDDAGNCTM-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Ti+4].[Co++]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results