accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1201844-73-6

IUPAC Name: 4-chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Synonyms: 4-chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Molecular Weight (g/mol): 289.57
Molecular Formula: C15H17BClNO2
InChi Key: ANPKKHFSNJKOAM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C2N=CC=C(Cl)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results