accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ONHOOSCl

CAS RN 120210-48-2

IUPAC Name: 5-chloro-2-hydroxy-3-(thiophene-2-carbonyl)-1H-indole-1-carboxamide
Synonyms: tenidap
Molecular Weight (g/mol): 320.75
Molecular Formula: C14H9ClN2O3S
InChi Key: IZSFDUMVCVVWKW-UHFFFAOYSA-N
SMILES: NC(=O)N1C(O)=C(C(=O)C2=CC=CS2)C2=CC(Cl)=CC=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results