accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12022-99-0

IUPAC Name: (silanylideneferrio)silane
Synonyms: (silanylideneferrio)silane
Molecular Weight (g/mol): 116.05
Molecular Formula: FeH4Si2
InChi Key: UVUDQHYBXBVHAI-UHFFFAOYSA-N
SMILES: [SiH2]=[Fe]=[SiH2]
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)