accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12023-53-9

PFeFeFe

CAS RN 12023-53-9

IUPAC Name: triferriophosphane
Synonyms: triferriophosphane
Molecular Weight (g/mol): 198.51
Molecular Formula: Fe3P
InChi Key: KFSKMQQNHSLZRJ-UHFFFAOYSA-N
SMILES: [Fe]P([Fe])[Fe]
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)