accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2GeTe

CAS RN 12025-39-7

IUPAC Name: germanetellone
Synonyms: germanetellone
Molecular Weight (g/mol): 202.25
Molecular Formula: GeH2Te
InChi Key: SBMPISTWTKUJKL-UHFFFAOYSA-N
SMILES: [GeH2]=[Te]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results