accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12026-05-0

IUPAC Name: rubidium(1+) hydroxide
Synonyms: rubidium hydroxide|RbOH
Molecular Weight (g/mol): 102.48
Molecular Formula: HORb
InChi Key: CPRMKOQKXYSDML-UHFFFAOYSA-M
SMILES: [OH-].[Rb+]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)