accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHOHOHOHOHNaNaSn

CAS RN 12027-70-2

IUPAC Name: tin(4+) disodium hexahydroxide
Synonyms: sodium stannate
Molecular Weight (g/mol): 266.73
Molecular Formula: H6Na2O6Sn
InChi Key: PMPBLIIMRRPPEO-UHFFFAOYSA-H
SMILES: [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Sn+4]
Grade 
  • (5)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)