accessibility menu, dialog, popup

UserName

Viewing profile as user:
OIOOIOO

CAS RN 12029-98-0

IUPAC Name: iodyl iodate
Synonyms: iodine pentoxide diiodine pentaoxide
Molecular Weight (g/mol): 333.8
Molecular Formula: I2O5
InChi Key: BIZCJSDBWZTASZ-UHFFFAOYSA-N
SMILES: O=[I](=O)O[I](=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)