accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1203-24-3

IUPAC Name: 1-(2-chlorophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(2-chlorophenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 207.61
Molecular Formula: C10H6ClNO2
InChi Key: KPQOXMCRYWDRSB-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1N1C(=O)C=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results