accessibility menu, dialog, popup

UserName

Viewing profile as user:
IrOO

CAS RN 12030-49-8

IUPAC Name: methane
Synonyms: methane
Molecular Weight (g/mol): 16.04
Molecular Formula: CH4
InChi Key: VNWKTOKETHGBQD-UHFFFAOYSA-N
SMILES: C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)