accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12031-66-2

IUPAC Name: tantalum(5+) lithium(1+) trioxidandiide
Synonyms: lithium tantalate
Molecular Weight (g/mol): 235.88
Molecular Formula: LiO3Ta
InChi Key: JNQQEOHHHGGZCY-UHFFFAOYSA-N
SMILES: [Li+].[O--].[O--].[O--].[Ta+5]
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)