accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNCH3I

CAS RN 120317-69-3

IUPAC Name: 1,4-dimethyl-1H-1,2,4-triazol-4-ium iodide
Synonyms: 1,4-dimethyl-1,2,4-triazol-4-ium iodide
Molecular Weight (g/mol): 225.03
Molecular Formula: C4H8IN3
InChi Key: IMFBMMNQDMTHGG-UHFFFAOYSA-M
SMILES: [I-].CN1C=[N+](C)C=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results