accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOMgTi

CAS RN 12032-30-3

IUPAC Name: titanium(4+) magnesium(2+) trioxidandiide
Synonyms: titanium(4+) magnesium(2+) trioxidandiide
Molecular Weight (g/mol): 120.17
Molecular Formula: MgO3Ti
InChi Key: XSETZKVZGUWPFM-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Mg++].[Ti+4]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results