accessibility menu, dialog, popup

UserName

Viewing profile as user:
MnTe

CAS RN 12032-88-1

IUPAC Name: tellanylidenemanganese
Synonyms: manganese(II) telluride
Molecular Weight (g/mol): 182.54
Molecular Formula: MnTe
InChi Key: VMINMXIEZOMBRH-UHFFFAOYSA-N
SMILES: [Mn]=[Te]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results