accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12033-29-3

IUPAC Name: trisulfanylidenemolybdenum
Synonyms: molybdenum trisulfide
Molecular Weight (g/mol): 192.13
Molecular Formula: MoS3
InChi Key: TVWWSIKTCILRBF-UHFFFAOYSA-N
SMILES: S=[Mo](=S)=S
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results