accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaTe

CAS RN 12034-41-2

IUPAC Name: disodium tellanediide
Synonyms: sodium telluride
Molecular Weight (g/mol): 173.58
Molecular Formula: Na2Te
InChi Key: QZBKIBAHGRAIOX-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[Te--]
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results