accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12034-59-2

IUPAC Name: dioxoniobium
Synonyms: niobium dioxide
Molecular Weight (g/mol): 124.9
Molecular Formula: NbO2
InChi Key: HFLAMWCKUFHSAZ-UHFFFAOYSA-N
SMILES: O=[Nb]=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)