accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12034-77-4

IUPAC Name: diselanylideneniobium
Synonyms: diselanylideneniobium
Molecular Weight (g/mol): 250.85
Molecular Formula: NbSe2
InChi Key: CXRFFSKFQFGBOT-UHFFFAOYSA-N
SMILES: [Se]=[Nb]=[Se]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results