accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12035-72-2

IUPAC Name: disulfanylidenetrinickel
Synonyms: disulfanylidenetrinickel
Molecular Weight (g/mol): 240.2
Molecular Formula: Ni3S2
InChi Key: YGHCWPXPAHSSNA-UHFFFAOYSA-N
SMILES: S=[Ni][Ni][Ni]=S
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)