accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12036-21-4

IUPAC Name: dioxovanadium
Synonyms: vanadium dioxide vanadium(IV) oxide
Molecular Weight (g/mol): 82.94
Molecular Formula: O2V
InChi Key: GRUMUEUJTSXQOI-UHFFFAOYSA-N
SMILES: O=[V]=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)