accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12036-32-7

IUPAC Name: dipraseodymium(3+) trioxidandiide
Synonyms: praseodymium(III) oxide
Molecular Weight (g/mol): 329.81
Molecular Formula: O3Pr2
InChi Key: MMKQUGHLEMYQSG-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Pr+3].[Pr+3]
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)