man

Test

Viewing profile as user:
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Main Menu
Admin
Main Menu
ULS Inventory Manager
Products

CAS RN 1203648-40-1

CH3NH3COBr

CAS RN 1203648-40-1

IUPAC Name: 1-(3-bromophenyl)-3-(dimethylamino)prop-2-en-1-one
Synonyms: 1-(3-bromophenyl)-3-(dimethylamino)prop-2-en-1-one
Molecular Weight (g/mol): 254.13
Molecular Formula: C11H12BrNO
InChi Key: FBICCVMQKCGCST-UHFFFAOYSA-N
SMILES: CN(C)C=CC(=O)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results