CAS RN 12037-29-5
Praseodymium(III, IV) oxide, REacton™, 99.99% (REO)
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Praseodymium(III, IV) oxide, REacton™, 99.996% (REO)
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Praseodymium(III, IV) oxide, 99.5% (REO)
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Praseodymium(III, IV) oxide, 99% (REO)
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Praseodymium(III,IV) oxide, 99.9%, (trace metal basis)
Praseodymium(III, IV) oxide, 99.9% (REO)
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Praseodymium AA Standard, 1000 ppm Pr in 3% HNO3, Ricca Chemical
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Praseodymium ICP Standard, 10,000 ppm Pr in 5% HNO3, Ricca Chemical
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Praseodymium ICP Standard, 1000 ppm Pr in 3% HNO3, Ricca Chemical
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Sigma Aldrich Praseodymium(III,IV) oxide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.