accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12037-58-0

IUPAC Name: dioxotungsten
Synonyms: tungsten(IV) oxide
Molecular Weight (g/mol): 215.84
Molecular Formula: O2W
InChi Key: DZKDPOPGYFUOGI-UHFFFAOYSA-N
SMILES: O=[W]=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results