accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12038-21-0

IUPAC Name: disulfanylideneplatinum
Synonyms: platinum disulfide
Molecular Weight (g/mol): 259.2
Molecular Formula: PtS2
InChi Key: RKNFJIIYAUSTJA-UHFFFAOYSA-N
SMILES: S=[Pt]=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results