accessibility menu, dialog, popup

UserName

Viewing profile as user:
SReS

CAS RN 12038-63-0

IUPAC Name: disulfanylidenerhenium
Synonyms: rhenium disulfide
Molecular Weight (g/mol): 250.33
Molecular Formula: ReS2
InChi Key: USWJSZNKYVUTIE-UHFFFAOYSA-N
SMILES: S=[Re]=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results