accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12039-13-3

IUPAC Name: disulfanylidenetitanium
Synonyms: titanium disulfide
Molecular Weight (g/mol): 111.99
Molecular Formula: S2Ti
InChi Key: CFJRPNFOLVDFMJ-UHFFFAOYSA-N
SMILES: S=[Ti]=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results