accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12039-83-7

IUPAC Name: (silanylidenetitanio)silane
Synonyms: titanium disilicide
Molecular Weight (g/mol): 108.07
Molecular Formula: H4Si2Ti
InChi Key: WBWPCELMONEWER-UHFFFAOYSA-N
SMILES: [SiH2]=[Ti]=[SiH2]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results