accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1204-85-9

IUPAC Name: 4-(piperidin-1-yl)benzonitrile
Synonyms: 4-(piperidin-1-yl)benzonitrile
Molecular Weight (g/mol): 186.26
Molecular Formula: C12H14N2
InChi Key: ZEPXHFFGXQFUDP-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)N1CCCCC1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)