accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1204518-08-0

IUPAC Name: [3-(trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodanium trifluoromethanesulfonate
Synonyms: [3-(trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodanium triflate
Molecular Weight (g/mol): 540.26
Molecular Formula: C17H15F6IO3S
InChi Key: FKQZCCNYTGBVPK-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.CC1=CC(C)=C([I+]C2=CC=CC(=C2)C(F)(F)F)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results