accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOOOOOOOOOOOOOOFeFeFeFeFeFeFeFeFeFeFeFeBa

CAS RN 12047-11-9

IUPAC Name: dodecairon(3+) barium(2+) nonadecaoxidandiide
Synonyms: dodecairon(3+) barium(2+) nonadecaoxidandiide
Molecular Weight (g/mol): 1111.45
Molecular Formula: BaFe12O19
InChi Key: HPYIMVBXZPJVBV-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Ba++]
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)