accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOCaTi

CAS RN 12049-50-2

IUPAC Name: titanium(4+) calcium trioxidandiide
Synonyms: calcium titanate
Molecular Weight (g/mol): 135.94
Molecular Formula: CaO3Ti
InChi Key: WEUCVIBPSSMHJG-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Ca++].[Ti+4]
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)