accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1205-06-7

IUPAC Name: ethyl 4-isothiocyanatobenzoate
Synonyms: ethyl 4-isothiocyanatobenzoate
Molecular Weight (g/mol): 207.25
Molecular Formula: C10H9NO2S
InChi Key: MLOJHUCMCKBDLV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)