accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12054-48-7

IUPAC Name: nickel(2+) dihydroxide
Synonyms: nickel(II) hydroxide
Molecular Weight (g/mol): 92.71
Molecular Formula: H2NiO2
InChi Key: BFDHFSHZJLFAMC-UHFFFAOYSA-L
SMILES: [OH-].[OH-].[Ni++]
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)