accessibility menu, dialog, popup

UserName

Viewing profile as user:
OLiLi

CAS RN 12057-24-8

IUPAC Name: (lithiooxy)lithium
Synonyms: —
Molecular Weight (g/mol): 29.88
Molecular Formula: Li2O
InChi Key: XZIUWXMPQBQHMC-UHFFFAOYSA-N
SMILES: [Li]O[Li]
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)