accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1206-46-8

IUPAC Name: trimethyl(2,3,4,5,6-pentafluorophenyl)silane
Synonyms: trimethyl(2,3,4,5,6-pentafluorophenyl)silane
Molecular Weight (g/mol): 240.25
Molecular Formula: C9H9F5Si
InChi Key: GABHTFORECKGBB-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results