accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12060-00-3

IUPAC Name: λ²-lead(2+) oxotitaniumbis(olate)
Synonyms: lead titanate
Molecular Weight (g/mol): 303.1
Molecular Formula: O3PbTi
InChi Key: NKZSPGSOXYXWQA-UHFFFAOYSA-N
SMILES: [Pb++].[O-][Ti]([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)