accessibility menu, dialog, popup

UserName

Viewing profile as user:
OZrOOPb

CAS RN 12060-01-4

IUPAC Name: λ²-lead(2+) oxozirconiumbis(olate)
Synonyms: λ²-lead(2+) oxozirconiumbis(olate)
Molecular Weight (g/mol): 346.4
Molecular Formula: O3PbZr
InChi Key: RSCSNKLWUCUPDE-UHFFFAOYSA-N
SMILES: [Pb++].[O-][Zr]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results