CAS RN 1206487-81-1
IUPAC Name:
11-benzyl-22-(cyclopropylmethyl)-14-oxa-11,22-diazaheptacyclo[13.9.1.0¹,¹³.0²,²¹.0⁴,¹².0⁵,¹⁰.0¹⁹,²⁵]pentacosa-4(12),5,7,9,15,17,19(25)-heptaene-2,16-diol hydrochloride
Synonyms:
11-benzyl-22-(cyclopropylmethyl)-14-oxa-11,22-diazaheptacyclo[13.9.1.0¹,¹³.0²,²¹.0⁴,¹².0⁵,¹⁰.0¹⁹,²⁵]pentacosa-4(12),5,7,9,15,17,19(25)-heptaene-2,16-diol hydrochloride
Molecular Weight (g/mol):
541.09
Molecular Formula:
C33H33ClN2O3
InChi Key:
KDEBSUYWJUBPCB-UHFFFAOYNA-N
SMILES:
Cl.OC1=C2OC3C4=C(CC5(O)C6CC(C=C1)=C2C35CCN6CC1CC1)C1=CC=CC=C1N4CC1=CC=CC=C1
1
–
1
of
1
results
1
N-Benzylnaltrindole hydrochloride, Tocris Bioscience™
CAS: 1206487-81-1 Molecular Formula: C33H33ClN2O3 Molecular Weight (g/mol): 541.088 InChI Key: KDEBSUYWJUBPCB-REBVNRAYSA-N Synonym: n-benzylnaltrindole hydrochloride PubChem CID: 131845649 SMILES: C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=CC=CC=C8N6CC9=CC=CC=C9.Cl