accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120659-34-9

S

CAS RN 120659-34-9

IUPAC Name: 3-cyclohexylthiophene
Synonyms: 3-cyclohexylthiophene
Molecular Weight (g/mol): 166.28
Molecular Formula: C10H14S
InChi Key: FGHYZEFIPLFAOC-UHFFFAOYSA-N
SMILES: C1CCC(CC1)C1=CSC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results