accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12067-46-8

IUPAC Name: diselanylidenetungsten
Synonyms: tungsten diselenide
Molecular Weight (g/mol): 341.78
Molecular Formula: Se2W
InChi Key: ROUIDRHELGULJS-UHFFFAOYSA-N
SMILES: [Se]=[W]=[Se]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)