CAS RN 12068-51-8
IUPAC Name:
dialuminium(3+) magnesium(2+) tetraoxidandiide
Synonyms:
dialuminium(3+) magnesium(2+) tetraoxidandiide
Molecular Weight (g/mol):
142.26
Molecular Formula:
Al2MgO4
InChi Key:
UAMZXLIURMNTHD-UHFFFAOYSA-N
SMILES:
[O--].[O--].[O--].[O--].[Mg++].[Al+3].[Al+3]
1
–
2
of
2
results