accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12068-69-8

IUPAC Name: dibismuth(3+) triselandiide
Synonyms: bismuth selenide
Molecular Weight (g/mol): 654.87
Molecular Formula: Bi2Se3
InChi Key: FBGGJHZVZAAUKJ-UHFFFAOYSA-N
SMILES: [Se--].[Se--].[Se--].[Bi+3].[Bi+3]
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)