accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12069-85-1

IUPAC Name: methanidylidynehafniumylium
Synonyms: hafnium(IV) carbide
Molecular Weight (g/mol): 190.5
Molecular Formula: CHf
InChi Key: NVDNLVYQHRUYJA-UHFFFAOYSA-N
SMILES: [C-]#[Hf+]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results