accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12069-94-2

IUPAC Name: methanidylidyneniobiumylium
Synonyms: methanidylidyneniobiumylium
Molecular Weight (g/mol): 104.92
Molecular Formula: CNb
InChi Key: AADPBXLVMHIYCT-UHFFFAOYSA-N
SMILES: [C-]#[Nb+]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results